Product Details
- SNP ID
-
rs145436915
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:19284772 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTGTCCTCATACTTGTACTTGCC[A/G]GTCAGCAGGCCCCCTGAGGGAAAGC
- Phenotype
-
MIM: 608477
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKR7A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1738023] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKR7A3
- Gene Name
- aldo-keto reductase family 7 member A3
- Gene
- LOC100506730
- Gene Name
- uncharacterized LOC100506730
There are no transcripts associated with this gene.
View Full Product Details