Product Details

SNP ID
rs145517238
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:111440356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTTAAAACCTTAGAAGTCTA[T/G]AAATACTGCATTCAGACTTTCCACA
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
There are no transcripts associated with this gene.

Gene
WDR77
Gene Name
WD repeat domain 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317062.1 2078 UTR 3 NP_001303991.1
NM_001317063.1 2078 UTR 3 NP_001303992.1
NM_001317064.1 2078 UTR 3 NP_001303993.1
NM_024102.3 2078 UTR 3 NP_077007.1

View Full Product Details