Product Details

SNP ID
rs146598146
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43653273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAACTTCAGTAGATACATTGCCT[A/G]CATTGAATCCCAAGGAGCTCATCGG
Phenotype
MIM: 609764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM4A PubMed Links

Gene Details

Gene
KDM4A
Gene Name
lysine demethylase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014663.2 301 Missense Mutation TAC,TGC Y33C NP_055478.2
XM_005271354.3 301 Missense Mutation TAC,TGC Y33C XP_005271411.1
XM_005271355.3 301 Missense Mutation TAC,TGC Y33C XP_005271412.1
XM_005271356.3 301 UTR 5 XP_005271413.1
XM_011542467.2 301 Intron XP_011540769.2
XM_017002912.1 301 UTR 5 XP_016858401.1

View Full Product Details