Product Details

SNP ID
rs147658835
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:208025104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAGTGGCCTTGGCATGGCTG[G/A]TTGTGAATTATAATGAAAACAGCAA
Phenotype
MIM: 601054
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PLXNA2 PubMed Links

Gene Details

Gene
PLXNA2
Gene Name
plexin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025179.3 8841 UTR 3 NP_079455.3
XM_005273164.2 8841 UTR 3 XP_005273221.1
XM_005273165.3 8841 Intron XP_005273222.1

View Full Product Details