Product Details

SNP ID
rs147810658
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:965185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACGCTGTCCGTGTCCTCCACCAG[C/T]CTCTGACCCACCTACCACCAGAGGC
Phenotype
MIM: 610770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198317.2 2583 Silent Mutation AGC,AGT S641S NP_938073.1
XM_006710600.3 2583 Silent Mutation AGC,AGT S664S XP_006710663.1
XM_006710601.3 2583 Intron XP_006710664.1
Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
There are no transcripts associated with this gene.

Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160184.1 2583 Intron NP_001153656.1
NM_032129.2 2583 Intron NP_115505.2
XM_006710944.3 2583 Intron XP_006711007.2
XM_011542248.2 2583 Intron XP_011540550.2
XM_017002474.1 2583 Intron XP_016857963.1
XM_017002475.1 2583 Intron XP_016857964.1
XM_017002476.1 2583 Intron XP_016857965.1
XM_017002477.1 2583 Intron XP_016857966.1
XM_017002478.1 2583 Intron XP_016857967.1
XM_017002479.1 2583 Intron XP_016857968.1

View Full Product Details