Product Details
- SNP ID
-
rs147905114
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:95476790 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACACGCACCCACATGAACACACA[C/T]GGGCACAAAGAGATACTACAGAATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ31662
PubMed Links
Gene Details
- Gene
- FLJ31662
- Gene Name
- uncharacterized LOC440594
There are no transcripts associated with this gene.
View Full Product Details