Product Details

SNP ID
rs148190419
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:37537964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCCAAGGTCAATGATGTAGGG[C/T]TTCACTCTTCGGCCAACTGTGCCAT
Phenotype
MIM: 608241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 1090 Silent Mutation AAA,AAG K325K NP_078976.2

View Full Product Details