Product Details

SNP ID
rs148298575
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:15411869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCCCAGGAAGGGGCGTTTTGC[A/C]ACTGCACTGTTTGAGAAAGGCTCTG
Phenotype
MIM: 616450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EFHD2 PubMed Links

Gene Details

Gene
EFHD2
Gene Name
EF-hand domain family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024329.5 Intron NP_077305.2
XM_005246000.3 Intron XP_005246057.1
Gene
LOC101927441
Gene Name
uncharacterized LOC101927441
There are no transcripts associated with this gene.

View Full Product Details