Product Details

SNP ID
rs149111792
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:89053380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTTATGGTACATGCCTTTCGT[C/T]GTCTCATTTTCGTCTGGAGATCCTG
Phenotype
MIM: 600411
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBP1 PubMed Links

Gene Details

Gene
GBP1
Gene Name
guanylate binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002053.2 Intron NP_002044.2

View Full Product Details