Product Details
- SNP ID
-
rs150145103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:12859978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAGCTGGTCAATTATCTAACGCC[A/T]ATTAAATATCTCAGAAAGTCATTGA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRAMEF2
PubMed Links
Gene Details
- Gene
- PRAMEF2
- Gene Name
- PRAME family member 2
View Full Product Details