Product Details

SNP ID
rs150416355
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:186444039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGACATGTACCTCTCTTTCAGG[C/T]AGTAACCCATATTCATTTAGGAAAG
Phenotype
MIM: 171490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724919 PubMed Links

Gene Details

Gene
LOC102724919
Gene Name
uncharacterized LOC102724919
There are no transcripts associated with this gene.

Gene
PDC
Gene Name
phosducin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002597.4 755 Silent Mutation CTA,CTG L227L NP_002588.3
NM_022576.3 755 Silent Mutation CTA,CTG L175L NP_072098.1
XM_011509603.2 755 Silent Mutation CTA,CTG L227L XP_011507905.1

View Full Product Details