Product Details

SNP ID
rs150740046
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:37540157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTCTTCCTCCATGTTACTTAC[A/C]GATATTGAAAGACCGCATGCTGCTT
Phenotype
MIM: 608241
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNIP1 PubMed Links

Gene Details

Gene
SNIP1
Gene Name
Smad nuclear interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024700.3 1041 Missense Mutation CGG,CTG R309L NP_078976.2

View Full Product Details