Product Details
- SNP ID
-
rs150886014
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:52602544 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTGTCGCTGGAGAAGTACCGCGG[A/C]TCGGTGAGTGCGCGGGGTCTGGCGG
- Phenotype
-
MIM: 615784
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GPX7
PubMed Links
Gene Details
- Gene
- GPX7
- Gene Name
- glutathione peroxidase 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015696.4 |
174 |
Silent Mutation |
GGA,GGC |
G45G |
NP_056511.2 |
View Full Product Details