Product Details

SNP ID
rs151026064
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:225488062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAACAAACAAACAAACAAACA[C/T]CACCAAAGCTTTCCGCCATTCATGC
Phenotype
MIM: 609061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENAH PubMed Links

Gene Details

Gene
ENAH
Gene Name
enabled homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008493.1 11708 UTR 3 NP_001008493.1
NM_018212.4 11708 UTR 3 NP_060682.2
XM_005273187.2 11708 UTR 3 XP_005273244.1
XM_011544229.2 11708 Intron XP_011542531.2
XM_017001746.1 11708 UTR 3 XP_016857235.1
XM_017001747.1 11708 Intron XP_016857236.1
XM_017001748.1 11708 Intron XP_016857237.1
XM_017001749.1 11708 Intron XP_016857238.1
XM_017001750.1 11708 Intron XP_016857239.1
XM_017001751.1 11708 UTR 3 XP_016857240.1
XM_017001752.1 11708 Intron XP_016857241.1
XM_017001753.1 11708 UTR 3 XP_016857242.1
XM_017001754.1 11708 UTR 3 XP_016857243.1
XM_017001755.1 11708 UTR 3 XP_016857244.1
XM_017001756.1 11708 UTR 3 XP_016857245.1
XM_017001757.1 11708 UTR 3 XP_016857246.1

View Full Product Details