Product Details

SNP ID
rs141708025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63940160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGGCAGCCTCACCAATGCCTGG[A/G]ATGATGCGGAAAAGGTCATTGACCC
Phenotype
MIM: 611203 MIM: 610866
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5 PubMed Links

Gene Details

Gene
DNAJC5
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
There are no transcripts associated with this gene.

Gene
MIR1914
Gene Name
microRNA 1914
There are no transcripts associated with this gene.

Gene
MIR647
Gene Name
microRNA 647
There are no transcripts associated with this gene.

Gene
UCKL1
Gene Name
uridine-cytidine kinase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193379.1 1838 Silent Mutation NP_001180308.1
NM_017859.3 1838 Silent Mutation NP_060329.2
XM_005260216.2 1838 Silent Mutation XP_005260273.1
XM_006723806.2 1838 Silent Mutation XP_006723869.1
XM_006723807.2 1838 Silent Mutation XP_006723870.1
XM_006723808.1 1838 Silent Mutation XP_006723871.1
XM_006723809.1 1838 Silent Mutation XP_006723872.1
XM_006723810.1 1838 Silent Mutation XP_006723873.1
XM_006723811.3 1838 Silent Mutation XP_006723874.1
XM_011528868.2 1838 Silent Mutation XP_011527170.1
XM_011528869.1 1838 Silent Mutation XP_011527171.1
XM_011528870.1 1838 Silent Mutation XP_011527172.1
XM_011528871.1 1838 Silent Mutation XP_011527173.1
XM_011528872.1 1838 Silent Mutation XP_011527174.1
XM_011528873.2 1838 Intron XP_011527175.1
XM_017027894.1 1838 Intron XP_016883383.1
XM_017027895.1 1838 Intron XP_016883384.1
XM_017027896.1 1838 Silent Mutation XP_016883385.1
XM_017027897.1 1838 Intron XP_016883386.1
XM_017027898.1 1838 Silent Mutation XP_016883387.1
XM_017027899.1 1838 Missense Mutation XP_016883388.1
XM_017027900.1 1838 Silent Mutation XP_016883389.1
XM_017027901.1 1838 Silent Mutation XP_016883390.1
XM_017027902.1 1838 Silent Mutation XP_016883391.1
XM_017027903.1 1838 Silent Mutation XP_016883392.1
XM_017027904.1 1838 Silent Mutation XP_016883393.1
XM_017027905.1 1838 Silent Mutation XP_016883394.1
XM_017027906.1 1838 Silent Mutation XP_016883395.1
XM_017027907.1 1838 Silent Mutation XP_016883396.1
XM_017027908.1 1838 Silent Mutation XP_016883397.1

View Full Product Details