Product Details
- SNP ID
-
rs144098855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44337171 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCATCCAGCTATGCCCCTGCTGC[C/T]CAGCACCGTGGGCCTGGCAGGCCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDML
PubMed Links
Gene Details
- Gene
- R3HDML
- Gene Name
- R3H domain containing like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178491.3 |
14 |
Missense Mutation |
CCC,CTC |
P5L |
NP_848586.1 |
View Full Product Details