Product Details
- SNP ID
-
rs148850942
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23603518 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAGCCTGCTGTGGGCTCACTTCC[C/T]TTTGTCCCTCGGAATGGCTTTGTCA
- Phenotype
-
MIM: 616543
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CST9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs726217] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CST9
- Gene Name
- cystatin 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008693.2 |
581 |
Missense Mutation |
AGG,GGG |
R158G |
NP_001008693.2 |
View Full Product Details