Product Details
- SNP ID
-
rs149889663
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:33008624 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTGCTGGCACTGCTGCTGCCCG[A/T]GGTCGGTGCCTCCACGCCAGGCACC
- Phenotype
-
MIM: 614108
MIM: 613942
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BPIFB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6088066] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BPIFB2
- Gene Name
- BPI fold containing family B member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025227.2 |
267 |
Missense Mutation |
GAG,GTG |
E17V |
NP_079503.1 |
- Gene
- SUN5
- Gene Name
- Sad1 and UNC84 domain containing 5
There are no transcripts associated with this gene.
View Full Product Details