Product Details

SNP ID
rs139428854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29008038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACTCTGTGGCATTCAGTATAC[A/G]AACATCTCCATGACAATGTTTTTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320724.1 640 Missense Mutation CGT,TGT R121C NP_001307653.1
NM_016940.2 640 Missense Mutation CGT,TGT R150C NP_058636.1

View Full Product Details