Product Details

SNP ID
rs143708359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30829654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATGGAAGTTGGTCCCATAGAT[A/G]GGGTATCCTGGGAAGTAGCTTCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP7-1 PubMed Links

Gene Details

Gene
KRTAP7-1
Gene Name
keratin associated protein 7-1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181606.2 79 Silent Mutation CCC,CCT P15P NP_853637.2

View Full Product Details