Product Details

SNP ID
rs144374953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37944447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTCAGCAAGGATGATTCCTCG[A/G]TACTGTTTGCCCTGTCCCAGGGCCT
Phenotype
MIM: 604414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf23 PubMed Links

Gene Details

Gene
C22orf23
Gene Name
chromosome 22 open reading frame 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207062.1 605 Silent Mutation TAC,TAT Y163Y NP_001193991.1
NM_032561.4 605 Silent Mutation TAC,TAT Y184Y NP_115950.3
XM_005261781.1 605 Silent Mutation TAC,TAT Y184Y XP_005261838.1
XM_005261782.3 605 Silent Mutation TAC,TAT Y184Y XP_005261839.1
XM_005261783.2 605 Silent Mutation TAC,TAT Y163Y XP_005261840.1
XM_005261784.2 605 Silent Mutation TAC,TAT Y163Y XP_005261841.1
Gene
MICALL1
Gene Name
MICAL like 1
There are no transcripts associated with this gene.

Gene
POLR2F
Gene Name
RNA polymerase II subunit F
There are no transcripts associated with this gene.

View Full Product Details