Product Details

SNP ID
rs145137288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37065500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCCAGTGAGGGTCTGGGCTGT[C/G]AGGGCCCAGGTGGCAGGCAGGGGTG
Phenotype
MIM: 616386 MIM: 609862
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD17 PubMed Links

Gene Details

Gene
KCTD17
Gene Name
potassium channel tetramerization domain containing 17
There are no transcripts associated with this gene.

Gene
TMPRSS6
Gene Name
transmembrane protease, serine 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289000.1 2823 UTR 3 NP_001275929.1
NM_001289001.1 2823 UTR 3 NP_001275930.1
NM_153609.3 2823 UTR 3 NP_705837.1
XM_011529989.2 2823 UTR 3 XP_011528291.1

View Full Product Details