Product Details

SNP ID
rs146229052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38085020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCAGAAAAGGGGGAAAGAGGT[T/C]AGGGGGCAAGAGGTGGGCCCCCCAG
Phenotype
MIM: 605926 MIM: 610409
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BAIAP2L2 PubMed Links

Gene Details

Gene
BAIAP2L2
Gene Name
BAI1 associated protein 2 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025045.5 2296 UTR 3 NP_079321.3
XM_005261751.4 2296 UTR 3 XP_005261808.1
XM_011530379.2 2296 UTR 3 XP_011528681.1
XM_011530380.2 2296 UTR 3 XP_011528682.1
XM_011530381.2 2296 UTR 3 XP_011528683.1
XM_011530382.2 2296 UTR 3 XP_011528684.1
XM_011530383.2 2296 UTR 3 XP_011528685.1
XM_011530384.2 2296 UTR 3 XP_011528686.1
XM_011530386.2 2296 UTR 3 XP_011528688.1
XM_011530387.2 2296 UTR 3 XP_011528689.1
XM_011530388.2 2296 UTR 3 XP_011528690.1
Gene
PICK1
Gene Name
protein interacting with PRKCA 1
There are no transcripts associated with this gene.

Gene
SLC16A8
Gene Name
solute carrier family 16 member 8
There are no transcripts associated with this gene.

View Full Product Details