Product Details

SNP ID
rs149954316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24014688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCATCCTAGCTGATTTTAGTTG[C/T]TTTTGCGGTATGAATTATGACTGAT
Phenotype
MIM: 604251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABIN1 PubMed Links

Gene Details

Gene
CABIN1
Gene Name
calcineurin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199281.1 Intron NP_001186210.1
NM_001201429.1 Intron NP_001188358.1
NM_012295.3 Intron NP_036427.1
XM_005261415.2 Intron XP_005261472.1
XM_005261416.2 Intron XP_005261473.1
XM_005261417.2 Intron XP_005261474.1
XM_005261419.2 Intron XP_005261476.1
XM_006724179.1 Intron XP_006724242.1
XM_006724180.1 Intron XP_006724243.1
XM_006724181.1 Intron XP_006724244.1
XM_011530026.1 Intron XP_011528328.1
XM_011530029.2 Intron XP_011528331.1
XM_011530030.2 Intron XP_011528332.1
XM_017028680.1 Intron XP_016884169.1
XM_017028681.1 Intron XP_016884170.1
XM_017028682.1 Intron XP_016884171.1
XM_017028683.1 Intron XP_016884172.1
XM_017028684.1 Intron XP_016884173.1
Gene
GSTTP1
Gene Name
glutathione S-transferase theta pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details