Product Details

SNP ID
rs138037221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171784286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGGTGGAAGAAAGGGCGAACCC[G/T]ATCATGAGTTATTTGCAATCAGTAG
Phenotype
MIM: 603667
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105373738 PubMed Links

Gene Details

Gene
LOC105373738
Gene Name
uncharacterized LOC105373738
There are no transcripts associated with this gene.

Gene
SLC25A12
Gene Name
solute carrier family 25 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003705.4 3117 UTR 3 NP_003696.2
XM_011512070.2 3117 Intron XP_011510372.1

View Full Product Details