Product Details

SNP ID
rs142885985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27082159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCAAGGAGCTGCAAGGCCTGCG[A/G]GGCGTCCTGCAAGGACTGAGCGGGC
Phenotype
MIM: 130660 MIM: 614058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMILIN1 PubMed Links

Gene Details

Gene
EMILIN1
Gene Name
elastin microfibril interfacer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007046.3 1087 Silent Mutation CGA,CGG R196R NP_008977.1
Gene
KHK
Gene Name
ketohexokinase
There are no transcripts associated with this gene.

View Full Product Details