Product Details

SNP ID
rs143269025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130191778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGAATCTTTAATTGCAAACAA[C/G]TTGAAAGCAGCCATCCTAGGGGTGG
Phenotype
MIM: 613450 MIM: 610457
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MZT2B PubMed Links

Gene Details

Gene
MZT2B
Gene Name
mitotic spindle organizing protein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005263795.4 Intron XP_005263852.1
XM_011511886.2 Intron XP_011510188.1
XM_011511887.2 Intron XP_011510189.1
XM_011511888.2 Intron XP_011510190.1
XM_011511889.2 Intron XP_011510191.1
XM_011511890.2 Intron XP_011510192.1
Gene
SMPD4
Gene Name
sphingomyelin phosphodiesterase 4
There are no transcripts associated with this gene.

Gene
TUBA3E
Gene Name
tubulin alpha 3e
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207312.2 Intron NP_997195.1

View Full Product Details