Product Details

SNP ID
rs145615349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3387779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCGAAGAGAACGAGACCGCATC[A/G]GAAGGCTCGAGTCCTCTCGCGGACA
Phenotype
MIM: 614139 MIM: 608998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC12 PubMed Links

Gene Details

Gene
TRAPPC12
Gene Name
trafficking protein particle complex 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321102.1 792 Silent Mutation TCA,TCG S52S NP_001308031.1
NM_016030.5 792 Silent Mutation TCA,TCG S52S NP_057114.5
XM_011510350.2 792 Silent Mutation TCA,TCG S52S XP_011508652.1
XM_011510352.2 792 Silent Mutation TCA,TCG S52S XP_011508654.1
XM_011510353.2 792 Silent Mutation TCA,TCG S52S XP_011508655.1
XM_011510354.2 792 Silent Mutation TCA,TCG S52S XP_011508656.1
XM_011510355.2 792 Intron XP_011508657.1
XM_011510356.1 792 Intron XP_011508658.1
XM_017004257.1 792 Silent Mutation TCA,TCG S52S XP_016859746.1
Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
There are no transcripts associated with this gene.

View Full Product Details