Product Details

SNP ID
rs146822895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:18563953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCAAGGAAAAGAGCTTCGTCT[A/G]TCTCTAGACCCCAGCGTCGAAGGGT
Phenotype
MIM: 610526 MIM: 616796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NT5C1B PubMed Links

Gene Details

Gene
NT5C1B
Gene Name
5'-nucleotidase, cytosolic IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002006.2 1737 Missense Mutation ACA,ATA T559I NP_001002006.1
NM_001199086.1 1737 Missense Mutation ACA,ATA T542I NP_001186015.1
NM_001199087.1 1737 Missense Mutation ACA,ATA T576I NP_001186016.1
NM_001199088.1 1737 Missense Mutation ACA,ATA T561I NP_001186017.1
NM_033253.3 1737 Missense Mutation ACA,ATA T499I NP_150278.2
Gene
NT5C1B-RDH14
Gene Name
NT5C1B-RDH14 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199103.1 1737 Intron NP_001186032.1
NM_001199104.1 1737 Missense Mutation ACA,ATA T559I NP_001186033.1
Gene
RDH14
Gene Name
retinol dehydrogenase 14 (all-trans/9-cis/11-cis)
There are no transcripts associated with this gene.

View Full Product Details