Product Details

SNP ID
rs148505771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46543082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGGCGCGCTGATGCTCAAGTG[C/T]GTGGTGGTCGGCGACGGGGCGGTGG
Phenotype
MIM: 605857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1E2 PubMed Links

Gene Details

Gene
ATP6V1E2
Gene Name
ATPase H+ transporting V1 subunit E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318063.1 674 Intron NP_001304992.1
XM_005264633.2 674 Intron XP_005264690.1
XM_005264634.2 674 Intron XP_005264691.1
XM_011533148.2 674 Intron XP_011531450.1
XM_011533149.2 674 Intron XP_011531451.1
XM_011533150.2 674 Intron XP_011531452.1
XM_011533151.2 674 Intron XP_011531453.1
XM_011533152.2 674 Intron XP_011531454.1
XM_011533153.2 674 Intron XP_011531455.1
XM_017005225.1 674 Intron XP_016860714.1
XM_017005226.1 674 Intron XP_016860715.1
Gene
RHOQ
Gene Name
ras homolog family member Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012249.3 674 Silent Mutation TGC,TGT C12C NP_036381.2
XM_005264229.2 674 Silent Mutation TGC,TGT C12C XP_005264286.1
XM_011532726.2 674 Silent Mutation TGC,TGT C12C XP_011531028.1
XM_011532728.2 674 Intron XP_011531030.1
XM_017003716.1 674 Silent Mutation TGC,TGT C12C XP_016859205.1

View Full Product Details