Product Details

SNP ID
rs144232410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14493453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAGACCCAACTTGGCGAGAGA[C/T]CCCAGCTGTCACTCCAACTAGGGCA
Phenotype
MIM: 186854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIP2 PubMed Links

Gene Details

Gene
GRIP2
Gene Name
glutamate receptor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080423.3 3335 UTR 3 NP_001073892.3
XM_011534141.2 3335 UTR 3 XP_011532443.1
XM_011534142.2 3335 UTR 3 XP_011532444.1
XM_011534143.2 3335 UTR 3 XP_011532445.1
XM_011534145.2 3335 UTR 3 XP_011532447.1
XM_011534146.2 3335 UTR 3 XP_011532448.1
Gene
SLC6A6
Gene Name
solute carrier family 6 member 6
There are no transcripts associated with this gene.

View Full Product Details