Product Details

SNP ID
rs144881083
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9904097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCCACAAAGCCTTGGTGTGTG[C/T]GAGTCTGGCACTGTTCCCGCTGTTT
Phenotype
MIM: 614995 MIM: 616012
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
IL17RE PubMed Links

Gene Details

Gene
IL17RE
Gene Name
interleukin 17 receptor E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193380.1 283 Nonsense Mutation CGA,TGA R72* NP_001180309.1
NM_153480.1 283 Nonsense Mutation CGA,TGA R72* NP_705613.1
NM_153481.1 283 Intron NP_705614.1
NM_153483.2 283 Nonsense Mutation CGA,TGA R112* NP_705616.2
XM_006712975.1 283 Intron XP_006713038.1
XM_006712976.1 283 UTR 5 XP_006713039.1
XM_011533361.2 283 Intron XP_011531663.1
XM_011533362.2 283 Nonsense Mutation CGA,TGA R72* XP_011531664.1
XM_011533363.2 283 Nonsense Mutation CGA,TGA R72* XP_011531665.1
XM_011533364.2 283 Nonsense Mutation CGA,TGA R72* XP_011531666.1
Gene
JAGN1
Gene Name
jagunal homolog 1
There are no transcripts associated with this gene.

View Full Product Details