Product Details

SNP ID
rs149022031
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48223742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGGGGGACATTCTGCTCTGCT[C/T]TGGCTGGGCTTGGCCACGTGTTGTT
Phenotype
MIM: 600474
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CAMP PubMed Links

Gene Details

Gene
CAMP
Gene Name
cathelicidin antimicrobial peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004345.4 Intron NP_004336.3

View Full Product Details