Product Details

SNP ID
rs139356908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:80267036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCAGCTTCTCTGGGCAGCCAAG[C/G]AAGTGGCTTGGAGCAGAGCAGTTTC
Phenotype
MIM: 165190
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FGF5 PubMed Links

Gene Details

Gene
FGF5
Gene Name
fibroblast growth factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291812.1 449 Intron NP_001278741.1
NM_004464.3 449 Missense Mutation GCA,GGA A71G NP_004455.2
NM_033143.2 449 Missense Mutation GCA,GGA A71G NP_149134.1

View Full Product Details