Product Details

SNP ID
rs139927255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70198582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAATTCTTTTTTTTGGCAGGTG[A/G]TGCCCTATGTATTCTCCTTCAAAAT
Phenotype
MIM: 614843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ODAM PubMed Links

Gene Details

Gene
ODAM
Gene Name
odontogenic, ameloblast asssociated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017855.3 396 Missense Mutation ATG,GTG M127V NP_060325.3
XM_005265698.3 396 Missense Mutation ATG,GTG M144V XP_005265755.1
XM_006714245.3 396 Missense Mutation ATG,GTG M144V XP_006714308.1
XM_017008331.1 396 Missense Mutation ATG,GTG M113V XP_016863820.1

View Full Product Details