Product Details

SNP ID
rs143899697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:988526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGACCCTTGTTCAAATAAGAT[A/G]TCAACCCTGAGCGTCAGGTCAGGCC
Phenotype
MIM: 252800 MIM: 610130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDUA PubMed Links

Gene Details

Gene
IDUA
Gene Name
iduronidase, alpha-L-
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000203.4 2580 Intron NP_000194.2
XM_011513461.2 2580 Intron XP_011511763.1
XM_017008163.1 2580 Intron XP_016863652.1
Gene
SLC26A1
Gene Name
solute carrier family 26 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022042.3 2580 UTR 3 NP_071325.2
NM_134425.2 2580 Intron NP_602297.1
NM_213613.3 2580 UTR 3 NP_998778.1

View Full Product Details