Product Details

SNP ID
rs144117333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77159463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATCTTGCACTGAAACTTTTGTC[C/T]TGGCTGTCAATATTTTGGACAGGTT
Phenotype
MIM: 603203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNG2 PubMed Links

Gene Details

Gene
CCNG2
Gene Name
cyclin G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004354.2 678 Silent Mutation CTG,TTG L79L NP_004345.1
XM_011532398.1 678 Silent Mutation CTG,TTG L79L XP_011530700.1
XM_011532399.2 678 Silent Mutation CTG,TTG L79L XP_011530701.1

View Full Product Details