Product Details

SNP ID
rs144121892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:52595183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGCCTTTGGGCTATCGCTGAC[C/T]TCAGAATGGTCTCACATCTCCCACA
Phenotype
MIM: 612849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP46 PubMed Links

Gene Details

Gene
USP46
Gene Name
ubiquitin specific peptidase 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134223.1 3707 UTR 3 NP_001127695.1
NM_001286767.1 3707 UTR 3 NP_001273696.1
NM_001286768.1 3707 UTR 3 NP_001273697.1
NM_022832.3 3707 UTR 3 NP_073743.2
XM_017008557.1 3707 UTR 3 XP_016864046.1

View Full Product Details