Product Details

SNP ID
rs144697393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAGCAGGAGTACGTGGTGGTC[C/G]AGGCGGAGGAAGAGCTTACCCCCAG
Phenotype
MIM: 138297
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 856 Missense Mutation CAG,GAG Q172E NP_001968.3
XM_017007877.1 856 Intron XP_016863366.1

View Full Product Details