Product Details

SNP ID
rs145948847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158203505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAAATTCTGCTTGAAAATTAC[A/G]TGTTAAATTGATTCCTTCTCCATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM144 PubMed Links

Gene Details

Gene
TMEM144
Gene Name
transmembrane protein 144
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018342.4 Intron NP_060812.2
XM_005263110.1 Intron XP_005263167.1
XM_005263112.3 Intron XP_005263169.1
XM_006714254.1 Intron XP_006714317.1
XM_017008366.1 Intron XP_016863855.1
XM_017008367.1 Intron XP_016863856.1
XM_017008368.1 Intron XP_016863857.1

View Full Product Details