Product Details

SNP ID
rs146982667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:164197162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCCTCCTCGCCCTCCTCAT[A/C]TTCCACTACCTGAGCATCTTCATCA
Phenotype
MIM: 606877 MIM: 613331
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANP32C PubMed Links

Gene Details

Gene
ANP32C
Gene Name
acidic nuclear phosphoprotein 32 family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012403.1 Intron NP_036535.1
Gene
MARCH1
Gene Name
membrane associated ring-CH-type finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166373.1 Intron NP_001159845.1
NM_017923.3 Intron NP_060393.1
XM_011532055.2 Intron XP_011530357.1
XM_017008334.1 Intron XP_016863823.1
XM_017008335.1 Intron XP_016863824.1
XM_017008336.1 Intron XP_016863825.1

View Full Product Details