Product Details

SNP ID
rs147714674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:26481540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGGAGATGGAGCCTTCCTTCT[C/G]CATCAGCTCTGCTCCTTCTCTTCCT
Phenotype
MIM: 118444
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCKAR PubMed Links

Gene Details

Gene
CCKAR
Gene Name
cholecystokinin A receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000730.2 1538 UTR 3 NP_000721.1

View Full Product Details