Product Details

SNP ID
rs148210947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:83458474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATATTTTAGGGTTTTGCTTCT[GATA/-]GATCTGCTTAAAGAGAATCAAGTC
Phenotype
MIM: 611143 MIM: 606769 MIM: 611983
Polymorphism
GATA/-, Insertion/deletion
Allele Nomenclature
Literature Links
FAM175A PubMed Links

Gene Details

Gene
FAM175A
Gene Name
family with sequence similarity 175 member A
There are no transcripts associated with this gene.

Gene
HELQ
Gene Name
helicase, POLQ-like
There are no transcripts associated with this gene.

Gene
MRPS18C
Gene Name
mitochondrial ribosomal protein S18C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297767.1 Intron NP_001284696.1
NM_001297769.1 Intron NP_001284698.1
NM_001297770.1 Intron NP_001284699.1
NM_016067.3 Intron NP_057151.1

View Full Product Details