Product Details

SNP ID
rs149235069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55432612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAATCTTCATTTGAATGCTCAA[G/A]TTTCATGGGGCAGGGTGGGGGGCGG
Phenotype
MIM: 601851 MIM: 614726
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CLOCK PubMed Links

Gene Details

Gene
CLOCK
Gene Name
clock circadian regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267843.1 5356 UTR 3 NP_001254772.1
NM_004898.3 5356 UTR 3 NP_004889.1
XM_005265787.2 5356 UTR 3 XP_005265844.1
XM_011534409.2 5356 UTR 3 XP_011532711.1
XM_011534410.2 5356 UTR 3 XP_011532712.1
XM_011534411.2 5356 UTR 3 XP_011532713.1
XM_017008854.1 5356 UTR 3 XP_016864343.1
XM_017008855.1 5356 UTR 3 XP_016864344.1
Gene
TMEM165
Gene Name
transmembrane protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018475.4 5356 Intron NP_060945.2
XM_011534394.2 5356 Intron XP_011532696.1
XM_017008412.1 5356 Intron XP_016863901.1

View Full Product Details