Product Details

SNP ID
rs142083952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69366568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTGACGCAAGTTCTTTGCCT[A/G]GTGTGGTTTTTCCAACCCCTGGTGT
Phenotype
MIM: 603139 MIM: 600822
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AK6 PubMed Links

Gene Details

Gene
AK6
Gene Name
adenylate kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015891.1 385 Missense Mutation CCA,CTA P16L NP_001015891.1
NM_016283.4 385 Intron NP_057367.1
Gene
RAD17
Gene Name
RAD17 checkpoint clamp loader component
There are no transcripts associated with this gene.

Gene
TAF9
Gene Name
TATA-box binding protein associated factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015892.1 385 UTR 5 NP_001015892.1
NM_003187.4 385 Intron NP_003178.1

View Full Product Details