Product Details

SNP ID
rs150263604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36607245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTCATTTGCGAGGAGTCCTTTG[A/G]TTTCCAGTTACCTCCCCCACAGGCG
Phenotype
MIM: 600111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC1A3 PubMed Links

Gene Details

Gene
SLC1A3
Gene Name
solute carrier family 1 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166695.2 168 Intron NP_001160167.1
NM_001166696.2 168 Intron NP_001160168.1
NM_001289939.1 168 Intron NP_001276868.1
NM_001289940.1 168 Intron NP_001276869.1
NM_004172.4 168 Intron NP_004163.3
XM_005248342.2 168 UTR 5 XP_005248399.1
XM_011514084.2 168 Intron XP_011512386.1

View Full Product Details