Product Details

SNP ID
rs150357305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126600984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATGGCGTTGGAGGTCGGCGAT[A/G]TGGAAGATGGGCAGCTTTCCGACTC
Phenotype
MIM: 107323 MIM: 604924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH7A1 PubMed Links

Gene Details

Gene
ALDH7A1
Gene Name
aldehyde dehydrogenase 7 family member A1
There are no transcripts associated with this gene.

Gene
PHAX
Gene Name
phosphorylated adaptor for RNA export
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032177.3 Intron NP_115553.2

View Full Product Details