Product Details

SNP ID
rs141670836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49434199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGACAGACTGCAATAAAGAA[C/T]AACTACCCTTTATTTTCTCAAATAA
Phenotype
MIM: 609058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MUT PubMed Links

Gene Details

Gene
MUT
Gene Name
methylmalonyl-CoA mutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000255.3 Intron NP_000246.2
XM_005249143.3 Intron XP_005249200.1

View Full Product Details