Product Details

SNP ID
rs143795730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26107600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCTTTAGCCTTTCTCCCGCTC[C/T]TAACAGTTTTAGGAGTTGTCGCTCT
Phenotype
MIM: 142712 MIM: 602827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H1T PubMed Links

Gene Details

Gene
HIST1H1T
Gene Name
histone cluster 1, H1t
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005323.3 537 Missense Mutation AAG,AGG K165R NP_005314.2
Gene
HIST1H4C
Gene Name
histone cluster 1, H4c
There are no transcripts associated with this gene.

View Full Product Details