Product Details
- SNP ID
-
rs145415328
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:154405992 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGTGTGAAAACAGCCACCAATTT[G/T]AGTCCCCCCAGCCCCTCAAAAAGGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CNKSR3
PubMed Links
Gene Details
- Gene
- CNKSR3
- Gene Name
- CNKSR family member 3
View Full Product Details